Learn More
Ergodyne™ Skullerz™ Loki Safety Glasses/Sunglasses
Designed to be worn as regular sunglasses or as safety goggles by removing the temples, attaching the included strap, and adding the removable foam gasket for a dust-beating seal.
$59.60 - $131.60
Specifications
Certifications/Compliance | ANSI Z87.1+, CSA Z94.3 and MIL-PRF 32432 Ballistic Impact |
---|---|
For Use With (Application) | Construction/Trades, Oil and Gas, Mining, Maintenance, Assembly/Fabrication, Manufacturing, Police/Tactical |
Catalog Number | Mfr. No. | Frame Color | Lens Tint | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Frame Color | Lens Tint | Price | Quantity & Availability | ||||
19-910-359
|
Ergodyne™
56000 |
Black | Clear |
Each for $59.60
|
|
||||
19-910-360
|
Ergodyne™
56003 |
Black | Clear with Fog-Off™ |
Each for $69.70
|
|
||||
19-910-362
|
Ergodyne™
56021 |
Black | Polarized copper |
Each for $127.50
|
|
||||
19-910-363
|
Ergodyne™
56030 |
Black | Smoke |
Each for $81.90
|
|
||||
19-910-364
|
Ergodyne™
56031 |
Black | Polarized smoke |
Each for $131.60
|
|
||||
19-910-365
|
Ergodyne™
56033 |
Black | Smoke with Fog-Off™ |
Each for $67.90
|
|
||||
19-910-368
|
Ergodyne™
56080 |
Black | Indoor/Outdoor |
Each for $59.60
|
|
||||
19-910-369
|
Ergodyne™
56083 |
Black | Indoor/Outdoor with Fog-Off™ |
Each for $67.90
|
|
||||
19-910-372
|
Ergodyne™
56103 |
Matte Gray | Clear with Fog-Off™ |
Each for $71.30
|
|
||||
19-910-373
|
Ergodyne™
56121 |
Matte Gray | Polarized copper |
Each for $131.60
|
|
||||
19-910-375
|
Ergodyne™
56131 |
Matte Gray | Polarized smoke |
Each for $127.50
|
|
||||
19-910-376
|
Ergodyne™
56133 |
Matte Gray | Smoke with Fog-Off™ |
Each for $69.70
|
|
||||
19-910-361
|
Ergodyne™
56020 |
Black | Copper | N/A | N/A | N/A | |||
19-910-366
|
Ergodyne™
56042 |
Black | Silver, Mirror | N/A | N/A | N/A | |||
19-910-367
|
Ergodyne™
56050 |
Black | Yellow | N/A | N/A | N/A | |||
19-910-370
|
Ergodyne™
56092 |
Black | Blue, Mirror | N/A | N/A | N/A | |||
19-910-371
|
Ergodyne™
56100 |
Matte Gray | Clear | N/A | N/A | N/A | |||
19-910-374
|
Ergodyne™
56130 |
Matte Gray | Smoke | N/A | N/A | N/A | |||
19-910-377
|
Ergodyne™
56150 |
Matte Gray | Yellow | N/A | N/A | N/A | |||
19-910-378
|
Ergodyne™
56180 |
Matte Gray | Indoor/Outdoor | N/A | N/A | N/A | |||
Description
- Protective work safety sunglasses feature tough, scratch-resistant lenses that filter 99.9% of harmful UVA, UVB and UVC rays to keep eyes safe in the sun.
- Durable frames bend and flex without breaking
- Non-slip rubber temples keep the glasses secure and resist sweat
- Available with polarized lenses or with anti-fogging solution
- Polarized glasses reduce glare and eye fatigue around water, snow and in other bright environments.
- Anti-glare glasses block the harsh reflected light from water, keeping eyes comfortable around glare.
- Fog-Off™ treatment won't wash off or wear off and is ideal for use with dust masks.
- Certifications and Testing:
- Impact-resistant safety glasses are tested to ANSI Z87.1+ High-Velocity Impact Protection and optical requirements
- Tested to MIL-PRF-32432 Ballistic Fragmentation, Class 1, which is the US Military Combat Eye Protection System Performance Specification for eyewear. To pass the military standard, safety glasses must withstand a steel projectile shot at 660 ft / 201m per second which is 4x faster than the Z87+ standard. Lenses must not puncture, crack or shatter. Frames must retain the lenses and not break.
- Ideal for protecting eyes during outdoor work like construction, carpentry/woodworking or landscaping or for outdoor activities like boating, skiing, fishing, hunting/shooting, and sports.
- Available in a variety of lens options ideally suited for a variety of tasks and light environments.
Specifications
ANSI Z87.1+, CSA Z94.3 and MIL-PRF 32432 Ballistic Impact | |
Construction/Trades, Oil and Gas, Mining, Maintenance, Assembly/Fabrication, Manufacturing, Police/Tactical |