Learn More
United Scientific™ Slide Tray, Cardboard
SDPSCI_ED
Protects your slides while holding and transferring them.
$323.00 - $566.00
Specifications
Material Cardboard
View More Specs
Products 2
Catalog Number Mfr. No. Slide Capacity Price Quantity & Availability  
Catalog Number Mfr. No. Slide Capacity Price Quantity & Availability  
S012577
SDPSCI_ED
View Documents
United Scientific™
62603
20
Pack of 50 for $323.00
missing translation for 'QuantityLeft'
Add to Cart
S012576
SDPSCI_ED
View Documents
United Scientific™
62604
30
Pack of 50 for $566.00
missing translation for 'QuantityLeft'
Add to Cart
Description
Description
  • Designed as a convenient solution for holding and transferring microscope slides. Fold-over cover helps protect the slides. The flat surface area on the inside of the cover is ideal for labeling or writing any necessary information. Recessed compartments protect the slides when the cover is closed.
  • Accommodates microscope slides in two sizes:
    • 75.0 x 25.0 x 0.8 to 1.2 mm
    • 76.0 x 26.0 x 0.8 to 1.2 mm
  • Recessed compartments to protect slides when closed
  • Heavy duty cardboard
  • Thumb cuts for easy removal of slides
  • Flat writing surface for specimen identification
Specifications
Specifications
Cardboard
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.