Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Slotted Masses
SCI_ED

Alternate for expensive brass weights.

Supplier:  Eisco™ PHZNW050

 View more versions of this product

Catalog No. S24443


missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart

Description

Description

  • Zinc die-casted with powder-coated
  • Accuracy ±2%
Specifications

Specifications

Powder-coated Slotted Mass Set
Zinc Die-cast
50 g
Accuracy ±2%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.