Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMER 28, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Positive regulator of autophagy in a mechanism independent from the mTOR pathway. Increases autophagosome synthesis and enhances clearance of model autophagy substrates such as mutant huntingtin and A53T α-synuclein, associated with Huntington's and Parkinson's diseases, respectively. Promotes reprogramming of fibroblasts to neural stem-like cells in combination with other chemical reagents.
Specifications
Specifications
Quantity | 10 mg |
Synonym | smer28, smer 28, n-allyl-6-bromoquinazolin-4-amine, 6-bromo-n-2-propenyl-4-quinazolinamine, 6-bromo-n-prop-2-en-1-yl quinazolin-4-amine, 3srb, cbmicro_021891, smer26, cambridge id 6164173, smer28 2 |
Formula Weight | 264.12 |
Percent Purity | >99% |
Chemical Name or Material | SMER 28 |
Safety and Handling
Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction