Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SN-6

Selective Na+/Ca2+ exchange inhibitor (reverse mode)

$257.00 - $1069.00

Specifications

Purity >99%
Product Type SN-6
Molecular Weight (g/mol) 402.16
Inhibitors SN-6
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
21-841-0
View Documents
Tocris Bioscience™
2184/10
10 mg
Each for $257.00
missing translation for 'QuantityLeft'
Add to Cart
 
21-845-0
View Documents
Tocris Bioscience™
2184/50
50 mg
Each for $1,069.00
missing translation for 'QuantityLeft'
Add to Cart
 
Description

Description

Selective Na+/Ca2+-exchange (NCX) inhibitor; displays some selectivity for NCX1. IC50 values are 2.9, 16 and 8.6 μM for inhibition of intracellular Na+-dependent 45Ca2+ uptake by cells expressing NCX1, NCX2 and NCX3 respectively. Has some affinity for mACh receptors (IC50 = 18 μM) but minimal activity against NCKX2 and various receptors and ion channels (IC50 > 30 μM). Preferentially blocks Ca2+ influx mode and is more selective for NCX isoforms than KB-R7943 (Cat. No. 1244). Anti-ischemic; potently protects against hypoxia-induced renal tubular cell damage (IC50 = 0.63 μM).
Specifications

Specifications

>99%
SN-6
402.16
SN-6
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.