Learn More
Innovative Research Inc Sngl Hmn Whl Bld Na Hep Unt
Supplier:  Innovative Research Inc IWB1NAHUNIT
Encompass_Preferred
Single Donor Human Whole Blood Na Heparin Unit - Single Donor Human Whole Blood from Innovative Research is collected from individual donors in an FDA-approved collection center. Whole blood must be held after draw for FDA-required viral testing. Rush shipments are available upon request (while test results are pending) - in the event the blood is shipped pending test results, any reactive results will be relayed once available. If you have a preference (viral tests pending vs. viral tests completed), please make a note.
Catalog No. 50-274-3946
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.