Learn More
Dunlop™ SNUGBOOT WorkPro Full Safety
Engineered for comfort for those who get the job done
Supplier: Dunlop™ NE68A93.11
Description
- SRC rated slip resistant outsole to conquer any terrain
- PUROTEX material creates the first breathable and waterproof upper
- Certified according to all major global safety standards (EN ISO 20345:2011 S5 CI CR SRC | CSA Z195-14 PR SD | ASTM F2413-18 M/I/C PR SD 100
Specifications
Black, Charcoal | |
11 | |
Standard | |
Full Safety |
Purofort™ and Purotex™ Upper | |
Men's | |
Yes |