Learn More
Dunlop SNUGBOOT WorkPro Full Safety
SDP
Engineered for comfort for those who get the job done
Supplier:  Dunlop NE68A93.06
 View more versions of this product
Catalog No. 18-500-557
Description
Description
  • SRC rated slip resistant outsole to conquer any terrain
  • PUROTEX material creates the first breathable and waterproof upper
  • Certified according to all major global safety standards (EN ISO 20345:2011 S5 CI CR SRC | CSA Z195-14 PR SD | ASTM F2413-18 M/I/C PR SD 100
Specifications
Specifications
Black, Charcoal
6
Standard
Full Safety
Purofort™ and Purotex™ Upper
Men's
Yes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.