Learn More
AG Scientific Inc SOC Medium Liquid 5x10 ML
Supplier:  AG Scientific Inc S28355X10ML
Encompass_Preferred
SOC Medium Liquid is a rich media used primarily in the recovery step of E. Coli competent cell transformations.
Catalog No. 50-226-8385
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.