Learn More
*PMW INC B. Braun #R5201-01 Irrigation Solution 0.9% Sodium Chloride, Preservative Free Not for Injection Bottle 500 ML SOD CHL, IRR SOL 0.9% 500ML (16/CS) BTL
Supplier:  *PMW INC 00264220110
Encompass_Preferred
PIC is a PVC-free, DEHP-free Plastic Irrigation Container, Bottle is designed for easy non-drip pouring, Bottle has a molded hand grip for easier container handling, The closure is sterile with recessed set port, Threaded screw cap covers for the sterile non-drip pouring lip, It is recommended that the product be stored at room temperature (25°C); however, brief exposure up to 40°C does not adversely affect the product
Catalog No. NC3261462
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.