Learn More
SOD1/Cu-Zn SOD Antibody (3E5), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | SOD1/Cu-Zn SOD |
---|---|
Clone | 3E5 |
Dilution | Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA |
Applications | Western Blot, Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
SOD1/Cu-Zn SOD Monoclonal antibody specifically detects SOD1/Cu-Zn SOD in Human samples. It is validated for Western Blot, Flow Cytometry, ELISASpecifications
SOD1/Cu-Zn SOD | |
Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ALS, ALS1, amyotrophic lateral sclerosis 1 (adult), Cu, Cu/Zn superoxide dismutase, EC 1.15.1.1, homodimer, hSod1, indophenoloxidase A, IPOA, SOD, SOD, soluble, superoxide dismutase [Cu-Zn], Superoxide dismutase 1, superoxide dismutase 1, soluble, Zn superoxide dismutase, EC 1.15.1.110superoxide dismutase, cystolic | |
A synthesized peptide derived from Human SOD1/Cu-Zn SOD [UniProt P00441] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
3E5 | |
Western Blot, Flow Cytometry, ELISA | |
Unconjugated | |
Rabbit | |
Biologically Active Proteins, Cancer, Diabetes Research, Mitochondrial Fusion Proteins, Neuronal Cell Markers, Neuroscience, Tumor Suppressors | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
6647 | |
IgG | |
Affinity purified |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.