Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ SOD1/Cu-Zn SOD Antibody Pair [HRP]

Polyclonal/Monoclonal Antibody Pair
$828.50
Specifications
Assay Range | 62.5 pg/mL |
---|---|
Assay Sensitivity | 62.5-4000 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Product Type | Antibody Pairs |
Description
This solid phase sandwich ELISA utilizes an antibody specific for Human SOD1/Cu-Zn SOD.Specifications
62.5 pg/mL | |
Sandwich ELISA | |
Antibody Pairs | |
Human | |
ALS, ALS1, amyotrophic lateral sclerosis 1 (adult), Cu, Cu/Zn superoxide dismutase, EC 1.15.1.1, homodimer, hSod1, indophenoloxidase A, IPOA, SOD, SOD, soluble, superoxide dismutase [Cu-Zn], Superoxide dismutase 1, superoxide dismutase 1, soluble, Zn superoxide dismutase, EC 1.15.1.110superoxide dismutase, cystolic | |
SOD1 | |
Biologically Active Proteins, Cancer, Diabetes Research, Mitochondrial Fusion Proteins, Neuronal Cell Markers, Neuroscience, Tumor Suppressors | |
SOD1/Cu-Zn SOD |
62.5-4000 pg/mL | |
HRP | |
This solid phase sandwich ELISA utilizes an antibody specific for Human SOD1/Cu-Zn SOD. | |
Sandwich ELISA | |
6647 | |
Rabbit/Mouse | |
RUO | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 4 ng/mL is recommended. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title