Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bisulfite ACS AR Granular, Macron Fine Chemicals™

Catalog No. p-3178131 Shop All Avantor Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
POLYSTORMOR
Poly

Chemical Identifiers

CAS 7631-90-5
Molecular Formula NaO3S
Molecular Weight (g/mol) 103.05
MDL Number MFCD00003530
InChI Key DWAQJAXMDSEUJJ-UHFFFAOYSA-L
Synonym sodium bisulfite, sodium hydrogen sulfite, sodium bisulphite, sodium hydrogensulfite, sodium sulfhydrate, monosodium sulfite, uantax sbs, sulfurous acid, monosodium salt, caswell no. 750, hydrogen sodium sulfite
PubChem CID 23665763
ChEBI CHEBI:26709
IUPAC Name sodium sulfite
SMILES [Na+].[O-]S([O-])=O

Specifications

Physical Form Granules
Quantity 500 g
Formula Weight 104.06
Grade ACS
Packaging POLYSTORMOR
Chemical Name or Material Sodium Bisulfite
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.