Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Catalog No. 02004045 Shop All Avantor Products
Click to view available options
Quantity:
Packaging:

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS

7647-14-5

Molecular Formula

ClNa

Molecular Weight (g/mol)

58.44

MDL Number

MFCD00003477

InChI Key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Synonym

sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex

PubChem CID

5234

ChEBI

CHEBI:26710

IUPAC Name

sodium chloride

SMILES

[Na+].[Cl-]

Specifications

Formula Weight

58.44

Physical Form

Crystals

Grade

Meets ACS reagent chemical requirements

Packaging

Poly bottle

Specific Gravity

1L = 2.16kg

Quantity

2.5kg

Chemical Name or Material

Sodium Chloride

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.