Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Diphenylamine Sulfonate Solution, Reagents
Click to view available options
Quantity:
500 mL
1 L
Specifications
Specifications
Chemical Name or Material | Sodium Diphenylamine Sulfonate Solution |
Packaging | Poly Bottle |
CAS | 7732-18-5, 7664-93-9, 7664-38-2 |
Quantity | 1 L |
Color | Green |
Synonym | Sodium Diphenylaminesulfonic Acid |
Grade | Reagent |
Boiling Point | 100°C |
Specific Gravity | 1.0121 |
Solubility | Completely miscible |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction