Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium dodecyl sulfate, >99%, MP Biomedicals™

Catalog No. ICN10291825 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:

Chemical Identifiers

CAS

151-21-3

Molecular Formula

C12H25NaO4S

Molecular Weight (g/mol)

288.38

MDL Number

MFCD00036175

InChI Key

DBMJMQXJHONAFJ-UHFFFAOYSA-M

Synonym

sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal

PubChem CID

3423265

ChEBI

CHEBI:8984

SMILES

[Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Color

White

Density

>1.1 at 20°C

Quantity

25 g

IUPAC Name

sodium dodecyl sulfate

Formula Weight

288.38

Percent Purity

>99%

Physical Form

Granular Powder

Chemical Name or Material

Sodium dodecyl sulfate

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.