Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Hydroxide, 10N Solution, Biotech Reagent, J.T.Baker™
Shop All Avantor Products
Click to view available options
Quantity:
1L
4L
19L
Packaging:
Narrow Mouth Polyethylene Bottle
Poly Bottle
Specifications
Specifications
Formula Weight | 40 |
Grade | Biotech Reagent |
Packaging | Narrow Mouth Polyethylene Bottle |
Specific Gravity | 1L = 1.32kg |
Quantity | 1L |
Chemical Name or Material | Sodium Hydroxide, 10N Solution |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction