Learn More
Electron Microscopy Sciences Sodium Hydroxide 2N Solution 225 ML
Carbonate-free. This solution is prepared from ACS Reagent grade sodium hydroxide, and is standardized colormetrically against potassium biphthalate primary reference material. Normal Concentration 1.98-2.02