Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Iodine Symporter Monoclonal Antibody (FP5), QED Bioscience™

Mouse Monoclonal Antibody
Supplier: Leinco Technologies Inc 11563100UG

This item is not returnable.
View return policy
Description
Sodium Iodine Symporter Monoclonal antibody specifically detects Sodium Iodine Symporter in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (PFA fixed), Western BlotSpecifications
Sodium Iodine Symporter | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 | |
Q63008, Q92911, Q99PN0 | |
Mouse | |
Protein A | |
RUO | |
114479, 114613, 6528 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (PFA fixed), Western Blot | |
FP5 | |
Unconjugated | |
SLC5A5 | |
Na(+)/I(-) cotransporter; Na(+)/I(-) symporter; Na(+)/I(-)-symporter; Na+/I- symporter; Nis; SLC5A5; sodium iodide symporter; Sodium/iodide cotransporter; sodium-iodide symporter; solute carrier family 5 (sodium iodide symporter), member 5; solute carrier family 5 (sodium/iodide cotransporter), member 5; solute carrier family 5 member 5; solute carrier family 5, member 5; TDH1 | |
Fusion protein consisting of aa468-643 of human NIS attached to mannose binding protein (MBP). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction