Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate Dibasic, MP Biomedicals™

Catalog No. MP21998025 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:

Highly hygroscopic and water soluble. Sodium Phosphate Dibasic, MP Biomedicals is a sodium salt of phosphoric acid. This white powder is an important component of running buffer for denaturing gel electrophoresis.

Sodium phosphate dibasic anhydrous

  • Used for preparing phosphate buffer, which in turn is utilized in X-gal staining
  • Induces glomerular calcification as well as other nephrotic syndrome in Sprague-Dawley rats

Chemical Identifiers

CAS

7558-79-4

Molecular Formula

HNa2O4P

Molecular Weight (g/mol)

141.96

MDL Number

MFCD00003496

InChI Key

BNIILDVGGAEEIG-UHFFFAOYSA-L

Synonym

disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt

PubChem CID

24203

ChEBI

CHEBI:34683

SMILES

[Na+].[Na+].OP([O-])([O-])=O

Specifications

Physical Form

Crystalline Powder

Quantity

500 g

Solubility Information

Freely soluble in water; insoluble in alcohol

IUPAC Name

disodium hydrogen phosphate

Formula Weight

141.98

Percent Purity

98.0 to 100.5%

Grade

USP

Recommended Storage

Room temperature, desiccate

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.