Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Potassium ATPase Alpha 3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP29923850UL
Description
Sodium Potassium ATPase Alpha 3 Polyclonal antibody specifically detects Sodium Potassium ATPase Alpha 3 in Mouse samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Sodium Potassium ATPase Alpha 3 | |
Polyclonal | |
Immunohistochemistry-Paraffin 1:200-1:1000 | |
alpha 3 polypeptide, ATPase, Na+/K+ transporting, alpha 3 polypeptide, dystonia 12, DYT12, EC 3.6.3, MGC13276, RDP, Sodium pump subunit alpha-3, sodium/potassium-transporting ATPase alpha-3 chain, sodium/potassium-transporting ATPase subunit alpha-3 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse Sodium Potassium ATPase Alpha 3. | |
50 μg | |
Primary | |
Mouse | |
Purified |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.0) | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
478 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction