Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Potassium ATPase Beta 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$231.00 - $262.00
Specifications
Antigen | Sodium Potassium ATPase Beta 1 |
---|---|
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry-Paraffin 1:1000-1:4000 |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Sodium Potassium ATPase Beta 1 Polyclonal antibody specifically detects Sodium Potassium ATPase Beta 1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
Sodium Potassium ATPase Beta 1 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cancer, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Plasma Membrane Markers, Signal Transduction | |
0.2 um filtered solution in PBS | |
481 | |
IgG | |
Antigen and protein A Affinity-purified |
Western Blot 1:500-1:2000, Immunohistochemistry-Paraffin 1:1000-1:4000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
adenosinetriphosphatase, ATP1B, ATPase, Na+/K+ transporting, beta 1 polypeptide, Beta 1-subunit of Na(+), K(+)-ATPase, MGC1798, Na, K-ATPase beta-1 polypeptide, sodium/potassium-dependent ATPase beta-1 subunit, Sodium/potassium-dependent ATPase subunit beta-1, sodium/potassium-transporting ATPase beta-1 chain, sodium/potassium-transporting ATPase subunit beta-1 | |
Produced in rabbits immunized with purified, recombinant Human Sodium Potassium ATPase Beta 1 (Accession#: P05026-1; Glu63-Ser303) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title