Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Potassium Tartrate, Tetrahydrate, Granular, FCC, 99-102%, Spectrum™ Chemical
SDP

Catalog No. 18606752 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
Packaging:

P1406, 6381-59-5, C4H4KNaO6•4H2O

Spectrum™ Chemical Sodium Potassium Tartrate, Tetrahydrate, Granular, FCC is used as a sequestrant and stabilizer. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS

6381-59-5

Molecular Formula

C4H12KNaO10

Molecular Weight (g/mol)

282.22

InChI Key

VZOPRCCTKLAGPN-ZFJVMAEJSA-L

IUPAC Name

potassium sodium (2R,3R)-2,3-dihydroxybutanedioate tetrahydrate

SMILES

O.O.O.O.[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Specifications

CAS Max %

1

Packaging

Fiber Drum

Quantity

50 kg

Percent Purity

99 to 102%

Grade

FCC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.