Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More

Fisher Science Education™ Sodium Silicate Solution, 37%

Quantity | 1 L |
---|---|
Packaging | 37% |
Chemical Identifiers
1344-09-8, 7732-18-5 | |
212.14 | |
ADPGKKZKGXANON-UHFFFAOYSA-N | |
23266 | |
disodium oxosilanebis(olate) pentahydrate |
H10Na2O8Si | |
MFCD00003492 | |
sodium metasilicate, sodium silicate, waterglass, water glass, sodium siliconate, sodium polysilicate, sodium sesquisilicate, sodium silicate glass, sodium silicate solution, sodium water glass | |
CHEBI:60720 | |
O.O.O.O.O.[Na+].[Na+].[O-][Si]([O-])=O |
Specifications
Colorless | |
37% w/v | |
MFCD00003492 | |
ADPGKKZKGXANON-UHFFFAOYSA-N | |
disodium oxosilanebis(olate) pentahydrate | |
23266 | |
37% | |
Solution |
1344-09-8, 7732-18-5 | |
H10Na2O8Si | |
sodium metasilicate, sodium silicate, waterglass, water glass, sodium siliconate, sodium polysilicate, sodium sesquisilicate, sodium silicate glass, sodium silicate solution, sodium water glass | |
O.O.O.O.O.[Na+].[Na+].[O-][Si]([O-])=O | |
212.14 | |
CHEBI:60720 | |
1 L | |
Sodium Silicate Solution, 37% |
Safety and Handling

ShelfLife : 5 years
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?