Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate, 0.0394 N For Iodine Adsorption Number of Carbon Black ASTM D1510, Reagents

Catalog No. p-200021885
Click to view available options
Quantity:

Sodium Thiosulfate, 0.0394 N For Iodine Adsorption Number of Carbon Black ASTM D1510, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS

7732-18-5, 10102-17-7, 71-41-0

Molecular Formula

H2O

Molecular Weight (g/mol)

18.02

InChI Key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Synonym

Sodium hyposulfite, Hypo, Hyposulfite of soda

IUPAC Name

water

SMILES

O

Specifications

Boiling Point

100°C

Color

Colorless

Physical Form

Liquid

Quantity

20 L

Grade

Reagent

Packaging

Cubitainer

Specific Gravity

1.005

Chemical Name or Material

Sodium Thiosulfate, 0.0394 N For Iodine Adsorption Number of Carbon Black ASTM D1510

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.