Learn More
Hach Company Sodium Thiosulfate Standard Solution, Stabilized, 0.0250 N, 1 L
Supplier:  Hach Company 2409353
Encompass_Preferred
For dissolved oxygen determination by Winkler titration. Stabilized sodium thiosulfate formulation
Catalog No. 50-199-5940
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.