Learn More
Hach Company Sodium Thiosulfate Standard Solution, Stabilized, 0.0109 N, 100 mL MDB
Supplier:  Hach Company 2408932
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Stabilized sodium thiosulfate titrant for dissolved oxygen determination
Catalog No. 50-199-5932
missing translation for 'QuantityLeft'