Learn More
Hach Company Sodium Thiosulfate Standard Solution, Stabilized, 0.0250 N, 3.78 L
Supplier:  Hach Company 2409317
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
For dissolved oxygen determination by Winkler titration. Stabilized sodium thiosulfate formulation
Catalog No. 50-199-5939
missing translation for 'QuantityLeft'