Learn More
Medchemexpress LLC HY-I0406 5mg Medchemexpress, Sofosbuvir impurity F CAS:1337482-17-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYI04065MG
Encompass_Preferred
Medchemexpress, HY-I0406 5mg Sofosbuvir impurity F CAS:1337482-17-3 Sofosbuvir impurity F, an diastereoisomer of sofosbuvir, is the impurity of sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3254
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.