Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

International Enviroguard™ Soft Scrubs™ Short Sleeve Shirts

Catalog No. p-7221825 Shop All International Enviroguard Systems Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large

Soft Scrubs™ Short Sleeve Shirts are disposable SMS scrubs that offer the comfort and safety you need to work long hours.

SMS Soft Scrub™ short sleeve shirts offer the comfort and safety you need to work long hours. These incredibly soft shirts offer modesty with complete ''no show through'' coverage. Shirts can easily be worn as a protective undergarment beneath a sterile gown. After each use, you can quickly and easily dispose of these shirts to help reduce the spread of germs, bioburdens, and other contagions. A round hemmed neck comfortably lays flat while a left chest pocket and right-front hip pocket allow you to store eyeglasses, small instruments, pens, and more while you're on the go. Possible use areas include pharmaceuticals, labs, healthcare, controlled environments, and more.

  • Anti-static fabric is made of an opaque material that doesn't show through and resists abrasions
  • Disposable to helps reduce the spread of germs, bioburdens, or contagions
  • Convenient, fabric loop for easily hanging up clothing
  • Left chest pocket and right-front hip pocket allow for storage of items such as pens, eyeglasses, and small instruments
  • Round hemmed neck comfortably lays flat

Specifications

Size Small
Gender Unisex
Material SMS
Color Blue
Neck Style Hemmed Crew
Seam Type Serged
Sleeve Style Short Sleeve
Disposable Single-use
Latex Free Yes
No. of Pockets 2
Product Type Scrub Shirt
Sterility Non-sterile
For Use With (Application) Pharmaceutical, Lab, Medical, Manufacturing
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.