Learn More
Cole-Parmer Cole-Parmer Copper (Cu+2) ISE Double junction solution kits
Supplier:  Cole-Parmer 2750265
Encompass_Preferred
These kits contain a 60 mL bottle of fill solution, 475 mL bottle of standard solution and 475 mL bottle of ISA.
Catalog No. 50-633-639
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.