Learn More
SOLVAY CHEMICALS INC Interox SG 35 Plus Biocide for Isolators 6x1L
Supplier: SOLVAY CHEMICALS INC 204967 6X1L
Interox SG-35 Plus is a biocide used in pharmaceutical packaging to prevent microbial contamination. It contains 35% hydrogen peroxide and is effective against bacteria, yeasts, molds, and viruses. The biocide can be directly incorporated into plastic resins and coatings at levels up to 1000 ppm. Key benefits include broad efficacy, compatibility with packaging materials, and low residue. Interox SG-35 Plus is an essential tool for pharmaceutical manufacturers looking to protect their products from microbial spoilage and contamination during storage and distribution.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.