Learn More
BURKLE INC Solvent pump footoperated 95cm
Supplier:  BURKLE INC 56033000
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Barrel pump for solvents with discharge hose and stopcock made of stainless steel/PTFE. Solvent pump foot operated for filling off high purity liquids such as solvents and flammable liquids.All medium-contacting parts are made of stainless steel AISI 304 (1.4301) and PTFE. The Burkle anti-static set can be used to ground the pump for dispensing flammable liquids so that no electrostatic charge forms.Due to the barrel screw joint and the stopcock, the container and the pump can be hermetically sealed. So that no dangerous vapors can escape after dispensing.The pump works according to the overpressure principle. An excess pressure is built up in the tank with the foot actuator, forcing the liquid out of the container. Easy foot operation of the pump with foot actuator leaves both hands free for dispensing. The air hose can be quickly and easily connected and disconnected with the practical clip connection.Available with discharge tube and also a smaller hand operated version.
Catalog No. NC2187878