Learn More
Epredia™ Solvent-Resistant White Biopsy Bags
Shop All Epredia Products- Minimize the risk of small specimen loss
- Reduce reagent carryover and pressure artifact often found with sponges
- Rapidly remove specimens with convenient peel-open bag
Specifications
Product Type
Biopsy Bag
Material
Solvent-Resistant Nylon
Color
White
Length (Metric)
31 mm
Width (Metric)
43 mm
Length (English)
1.2 in.
Width (English)
1.7 in.
Dimensions (L x W)
1.2 x 1.7 in.
Size
Small
Quantity
1000/Cs.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.