Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sonepiprazole, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective, high affinity antagonist for the rat and human dopamine D4 receptor (Ki = 10 nM). Exhibits low affinity at other monoamine receptors (Ki > 2000 nM). Induces c-fos gene expression in medial prefrontal cortex in a similar manner to clozapine (Cat. No. 0444). Brain penetrant and orally available.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 401.52 |
Percent Purity | >98% |
Chemical Name or Material | Sonepiprazole |
Safety and Handling
Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction