Learn More
Sonoco™ Thermosafe EPS Foam-Only Shipper Foam Rings
Increase the flexibility of your warehouse inventory by utilizing foam rings on standard shippers.
$107.00
Catalog Number | Mfr. No. | Applications | Quantity | Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Applications | Quantity | Size | Price | Quantity & Availability | |||
03-001-250
|
Sonoco™
807R3 |
ThermoSafe™ EPS insulated shipper having 13.25 L x 6.25 W x 1.875 in. H inner and 16.25 L x 9.25 W x 4.875 in. H outer dimension | 20 foam only shipper | 3 in. (7.62 cm) |
Pack of 24 for $107.00
|
|
|||
03-001-107
|
Sonoco™
808R3 |
ThermoSafe™ EPS insulated shipper having 12.5 L x 12.5 W x 6 in. H inner and 15.5 L x 15.5 W x 9 in. H outer dimension | 4 foam only shipper | 3 in. (7.62 cm) | N/A | N/A | N/A | ||
03-001-112
|
Sonoco™
489UPSR2 |
Multipurpose, dome style Foam shipper | 2 in. (5.08 cm) | N/A | N/A | N/A | |||
Description
- Patented components basic container (body and dome lid), and stackable rings enable you to just add rings to increase package height
- Create a perfect match between payload and container capacity
- Increase packing flexibility while decreasing packaging inventory
- Simplify ordering warehouse only one box footprint plus rings to vary capacity as needed
- Designed to maximize temperature control and withstand the hazards of transport
- Available for eight of our most popular size foam containers
Specifications
Foam Ring | |
Foam |