Learn More
Bel-Art™ SP Scienceware™ Polyethylene Utility Bags
SCI_ED
Reusable, transparent bags are well-suited for shipping or storage and are especially useful as sample bags
Supplier:  Bel-Art™ H131761012/EMD
 View more versions of this product
Catalog No. S42884
Description
Description
  • FDA approved material; Latex free
  • Different sizes available to best match particular packaging requirements
  • Not autoclavable
Specifications
Specifications
Polyethylene
12 x 10 in.
305 mm
254 mm
0.038 mm
12 in.
10 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.