Learn More
Biolegend Spark BlueTM 574 anti-human CD
Supplier:  Biolegend 344783
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Spark Blue™ 574 anti-human CD8 [SK1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-272-2231