Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ SPC™ Absorbents SpillFix Granular
Brady SpillFix granular is completely natural producing no carcinogens or dust. With more than four times the absorption power of clay granular, this product is perfect for both indoor and outdoor use.
Supplier: Brady™ SF7
This item is not returnable.
View return policy
Description
- Premium absorbent made of 100% recycled material
- Effective on the absorption of hydrocarbons, water-based fluids and most aggressive chemicals
- Certified safe for landfill disposal
Specifications
Biodegradable Granular | |
Absorbent | |
7 lb. bag |
Absorption of hydrocarbons, water-based fluids and most aggressive chemicals | |
OSHA 29 CFR 1910.22(a)(2) ; EPA 40 CFR 112.7(c)(1)(vii); OSHA 29 CFR 1910,120(j)(1)(vii) | |
Granular |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction