Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Speakman™ Eyesaver™ Wall-Mounted Eye/Face Washes

Wall-mounted emergency eyewash

$584.87

Specifications

Certifications/Compliance ANSI Z358.1
Flow Rate 3.6 gpm
Mounting Wall
Pressure 30 psi
Product Type Eyewash
View More Specs

Includes: Emergency sign

Products 1
Catalog Number Mfr. No. Activation Includes Price Quantity & Availability  
Catalog Number Mfr. No. Activation Includes Price Quantity & Availability  
19-003-209
View Documents
Speakman™
SE582
Push Handle Stainless Steel bowl, dual yellow ABS Plastic aerated spray head, automatic flow controls, flip-top dust covers, Chrome Plated valve
Each of 1 for $584.87
missing translation for 'QuantityLeft'
Add to Cart
 
Description

Description

  • Stainless steel bowl
  • Dual aerated spray outlets
  • Wall mount comes with bracket and chrome plated tailpiece
  • Comfortably flush eyes for the ANSI recommended 15 minutes

Specifications

Specifications

ANSI Z358.1
Wall
Eyewash
3.6 gpm
30 psi
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.