Learn More
Gold Biotechnology Inc Spectinomycin Dihydrochloride Pentahydrate, USP Grade 5 g
Supplier: Gold Biotechnology Inc S1405
Spectinomycin is an aminocyclitol antibiotic isolated from Streptomyces spectabilis. It is effective against gram-positive and gram-negative bacteria, but most commonly used against Gonococcus species. It has also been used to monitor cell fate during leaf development and as a selection marker in plants to display successful transformation of the Spcr gene.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.