Learn More
Enzo Life Sciences Spectinomycin . sulfate, (10g), CAS Number: 64058-48-6
SDP
Supplier:  Enzo Life Sciences BMLA2810010
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Aminocyclitol antibiotic. Antibiotic. Inhibits protein synthesis (elongation) by interfering with peptidyl tRNA translocation. Effective against Gram-negative and Gram-positive bacteria (Gonnococcus only). Mutation in rpsE (the gene for ribosomal protein S5) prevents binding of spectinomycin. Alternative Name: U-18409E. Formula: C14H24N2O7. H2SO4. MW: 430.4. CAS Number: 64058-48-6. Purity: ≥98% (HPLC). Appearance: White solid. Solubility: Soluble in water. Long Term Storage: -20°C
Catalog No. NC9820645
missing translation for 'QuantityLeft'