Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FyterTech Nonwovens Spilfyter™ Mercury Vapor Suppressors
SDP

Catalog No. 191300027
Click to view available options
:
Do not use Vapor Suppressor on powdered aluminum or strong oxidizers (e.g. ozone, liquid oxygen, chlorine, peroxide, permanganate, nitric acid and ketone) as contact may result in fire. Avoid contact with high concentrations of ketone (e.g. Acetone, Methyl Ethyl Ketone, ect.) in air or liquid. Do not use this product on ammonium and ammonia-containing compounds as contact will cause generation of gases
Do not use Vapor Suppressor on powdered aluminum or strong oxidizers (e.g. ozone, liquid oxygen, chlorine, peroxide, permanganate, nitric acid and ketone) as contact may result in fire. Avoid contact with high concentrations of ketone (e.g. Acetone, Methyl Ethyl Ketone, ect.) in air or liquid. Do not use this product on ammonium and ammonia-containing compounds as contact will cause generation of gases

Designed for the removal of mercury (Hg), which is difficult to remove by other absorbents, including standard activated carbon

  • Component: Activated Carbon
  • Odorless, black, irregular, dry granular solid

Specifications

Color

Black

Material

Activated Carbon

Type

Mercury Vapor Suppressor

Format

Granular

For Use With (Application)

For the removal of mercury which is difficult to remove by other absorbents, including standard activated carbon

Packaging Quantity

5/Cs.

Weight (English)

7 lb.

Weight (Metric)

3 kg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.