Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assembly, Divinylbenzene/Carboxen™/Polydimethylsiloxane (DVB/CAR/PDMS), df 50/30 μm
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS) is manufactured with a flexible metal alloy used in the needle, plunger, and fiber core.
Supplier:  MilliporeSigma™ Supelco™ FFA57298U
 View more versions of this product
Catalog No. 11-100-8021
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Description
Description
The new metal alloy design includes a thicker, flexible plunger that is much less likely to kink or break, and helps to reinforce the needle especially when used in an auto-sampler with a sample agitator. Since the needle is more flexible and has a thinner wall than the standard stainless steel needle, a bevel has been placed on the needle to help it pierce septa materials more easily.
Specifications
Specifications
SPME
Solid Phase Micro-Extraction (SPME) Fibers & Accessories
23 ga
Multi-Fiber Exchanger
Plain gray hub
3/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
Carboxen and StableFlex are registered trademarks of Sigma-Aldrich Co. LLC