Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assembly, Divinylbenzene/Carboxen™/Polydimethylsiloxane (DVB/CAR/PDMS), df 50/30 μm
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS) is manufactured with a flexible metal alloy used in the needle, plunger, and fiber core.
Supplier: MilliporeSigma™ Supelco™ FFA57329U
Description
The new metal alloy design includes a thicker, flexible plunger that is much less likely to kink or break, and helps to reinforce the needle especially when used in an auto-sampler with a sample agitator. Since the needle is more flexible and has a thinner wall than the standard stainless steel needle, a bevel has been placed on the needle to help it pierce septa materials more easily.Specifications
SPME | |
Solid Phase Micro-Extraction (SPME) Fibers & Accessories | |
24 ga |
Multi-Fiber Exchanger | |
StableFlex Fiber | |
3/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Carboxen and StableFlex are registered trademarks of Sigma-Aldrich Co. LLC
Spot an opportunity for improvement?Share a Content Correction