Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assembly, Polyacrylate (PA), df 85 μm
Solid phase microextraction technique can be used for GC/MS analysis. It helps in simplifying and speeds up the sample preparation. It also reduces solvent requirement for the extraction.
Supplier:  MilliporeSigma™ Supelco™ FFA57294U
 View more versions of this product
Catalog No. 11-100-8019
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Description
Description
Specifications
Specifications
Solid Phase Micro-Extraction (SPME) Fibers & Accessories
3/Pk.
SPME
Fused Silica Fiber
23 ga
Multi-Fiber Exchanger
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.