Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assembly, Polyacrylate (PA), df 85 μm
Solid phase microextraction technique can be used for GC/MS analysis. It helps in simplifying and speeds up the sample preparation. It also reduces solvent requirement for the extraction.
Supplier: MilliporeSigma™ Supelco™ FFA57305
Specifications
SPME | |
Solid Phase Micro-Extraction (SPME) Fibers & Accessories | |
24 ga |
Multi-Fiber Exchanger | |
Fused Silica Fiber | |
3/Pk. |