Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assortment Kit 5
SPME Fiber Assortment Kit 5 contains one fiber of each: 65 μm PDMS/DVB coating, 50/30 μm DVB/Carboxen/PDMS coating, 85 μm Carboxen/PDMS coating and 100 μm PDMS coating
Supplier: MilliporeSigma™ Supelco™ FFA57362U
Specifications
SPME | |
Solid Phase Micro-Extraction (SPME) Fibers & Accessories | |
23 ga |
Multi-Fiber Exchanger | |
Fused Silica Fiber |