Learn More
Andwin Scientific SPOR-KLENZ 4X1QT/CS
SDP
Supplier:  Andwin Scientific 21899861
Encompass
Made with a mixture of peracetic acid, hydrogen peroxide, and acetic acid, the Spor-Klenz Concentrated Sterilant/Disinfectant is a liquid sterilant/disinfectant that performs its task of disinfecting in quick 10 minutes. This product is ideal for procedures wherein a critically high-level microbial control is necessary. It is sold in a case of four 1 quart bottles.
Catalog No. NC9778613
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.