Learn More
Benchmark Products Inc SPOR-KLENZ RTU 4 X 750ML
SDP
Supplier:  Benchmark Products Inc C6525M24X750ML
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Spor-Klenz Ready-To-Use, 850 ML � 4/Case STERIS' Spor-Klenz Ready-To-Use Cold Sterilant is a fast acting, liquid cold sterilant/disinfectant, filtered to 0.22 micron and specifically formulated for use in the sterilization and disinfection of hard environmental surfaces in Pharmaceutical, Medical Device, Biotech and Cosmetic manufacturing facilities. This product is a stabilized blend of peracetic acid, hydrogen peroxide, and acetic that provides fast, effective microbial control, including spores. All of the active ingredients are notified according to the European Union Biocidal Products Directive (BPD). Spor-Klenz Ready-To-Use Sterilant offers a low toxicity profile, requires no mixing or activation and is reusable for up to 14 days which reduces waste. Each lot is fully traceable and documented by a Certificate of Analysis including filtration and USP sterility. 64-19-7
Catalog No. NC9285535